1,3-Difluor-4-iodbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,4-Difluoroiodobenzene, 2,4-Difluoro-1-iodobenzene
004818-25G
004818-100G
FLUO004818-25GEA
45.9
EUR
FLUO004818-25G
FLUO004818-100G
1,3-Difluor-4-iodbenzol
1,3-Difluor-4-iodbenzol
Formel:
C₆H₃F₂I Molecular Weight: 239,99 g/mol Siedepunkt: 175…176 °C (1013 hPa) Dichte: 2 g/cm³ (20 °C) |
MDL:
MFCD00040151 CAS-Nummer: 2265-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...