1,4-Diiodtetrafluorbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1,2,4,5-Tetrafluoro-3,6-diiodobenzene

010456-5G 010456-1G
FLUO010456-5GEA 365 EUR
FLUO010456-5G FLUO010456-1G
1,4-Diiodtetrafluorbenzol
1,4-Diiodtetrafluorbenzol
Formel: C₆F₄I₂
Molecular Weight: 401,87 g/mol
MDL: MFCD00001055
CAS-Nummer: 392-57-4
EINECS: 206-877-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR