2-Fluor-6-iodbenzoesäure

Lieferant: FLUOROCHEM
Achtung

006243-1G 006243-5G 006243-25G
FLUO006243-1GEA 31.1 EUR
FLUO006243-1G FLUO006243-5G FLUO006243-25G
2-Fluor-6-iodbenzoesäure
2-Fluoro-6-iodobenzoic acid
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
Siedepunkt: 312 °C (1013 hPa)
Schmelzpunkt: 123…126 °C
MDL: MFCD00042289
CAS-Nummer: 111771-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR