2,3,6-Trifluoranilin

Lieferant: FLUOROCHEM

001839-1G 001839-5G
FLUO001839-1GEA 48.8 EUR
FLUO001839-1G FLUO001839-5G
2,3,6-Trifluoranilin
2,3,6-Trifluoranilin
Formel: C₆H₄F₃N
Molecular Weight: 147,1 g/mol
Siedepunkt: 150…151 °C (1013 hPa)
Dichte: 1,39 g/cm³ (20 °C)
MDL: MFCD00012369
CAS-Nummer: 67815-56-9
UN: 2941
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR