2,3-Difluoranilin

Lieferant: FLUOROCHEM

001855-5G 001855-25G 001855-100G
FLUO001855-5GEA 40.2 EUR
FLUO001855-5G FLUO001855-25G FLUO001855-100G
2,3-Difluoranilin
2,3-Difluoranilin
Formel: C₆H₅F₂N
Molecular Weight: 129.11 g/mol
Siedepunkt: 69 °C (12 mmHg)
Dichte: 1.274 g/cm³ (20 °C)
MDL: MFCD00010298
CAS-Nummer: 4519-40-8
EINECS: 224-847-2
UN: 2941
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR