2,4,6-Trifluoranilin
Lieferant: FLUOROCHEM
001887-10G
001887-50G
FLUO001887-10GEA
67.1
EUR
FLUO001887-10G
FLUO001887-50G
2,4,6-Trifluoranilin
2,4,6-Trifluoranilin
Formel:
C₆H₄F₃N Molecular Weight: 147.1 g/mol Siedepunkt: 152 Schmelzpunkt: 33…34 °C |
MDL:
MFCD00007650 CAS-Nummer: 363-81-5 EINECS: 206-660-8 UN: 2926 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...