3,4,5-Trifluoranilin

Lieferant: FLUOROCHEM
Achtung

002064-5G 002064-25G
FLUO002064-5GEA 45.9 EUR
FLUO002064-5G FLUO002064-25G
3,4,5-Trifluoranilin
3,4,5-Trifluoranilin
Formel: C₆H₄F₃N
Molecular Weight: 147,1 g/mol
MDL: MFCD00083522
CAS-Nummer: 163733-96-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR