1-Fluor-4-nitrobenzol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Fluornitrobenzol
A11057.36
A11057.0E
A11057.22
A11057.36EA
188
EUR
A11057.36
A11057.0E
A11057.22
1-Fluor-4-nitrobenzol ≥99%
1-Fluor-4-nitrobenzol
Purity: 99%
CAS No.: 350-46-9
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Flash Point: 83°(181°F)
Refractive Index: 1.5315
Melting Point: 22-24°
Boiling Point: 204-206°
Beilstein Reference: 606923
UN No.: 2811
MDL No.: MFCD00007282
Formel:
C₆H₄FNO₂ Molecular Weight: 141,1 g/mol Siedepunkt: 204…206 °C Schmelzpunkt: 22…24 °C Dichte: 1,330 Flammpunkt: 83 °C (181 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007282 CAS-Nummer: 350-46-9 EINECS: 206-502-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...