1-Nitro-2,3,4,5-tetrafluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,3,4,5-Tetrafluoronitrobenzene, 1,2,3,4-Tetrafluoro-5-nitrobenzene
007368-1G
007368-5G
007368-25G
FLUO007368-1GEA
31.1
EUR
FLUO007368-1G
FLUO007368-5G
FLUO007368-25G
1-Nitro-2,3,4,5-tetrafluorbenzol
1-Nitro-2,3,4,5-tetrafluorbenzol
Formel:
C₆HF₄NO₂ Molecular Weight: 195,07 g/mol Siedepunkt: 179 °C (1013 hPa) Dichte: 1,61 g/cm³ (20 °C) |
MDL:
MFCD00014686 CAS-Nummer: 5580-79-0 EINECS: 226-972-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...