1,2,3-Trifluor-5-nitrobenzol 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,4,5-Trifluornitrobenzol

L20302.06 L20302.14
L20302.06EA 73.7 EUR
L20302.06 L20302.14
1,2,3-Trifluor-5-nitrobenzol 99%
1,2,3-Trifluor-5-nitrobenzol

Purity: 99% CAS No.: 66684-58-0 Molecular Formula: C6H2F3NO2 Formula Weight: 177.08 Flash Point: 75°(167°F) Refractive Index: 1.4860 Beilstein Reference: 2455018 UN No.: 2810 MDL No.: MFCD00456803

Formel: C₆H₂F₃NO₂
Molecular Weight: 177,08 g/mol
Dichte: 1,52
Flammpunkt: 75 °C (167 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00456803
CAS-Nummer: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR