Pentafluornitrobenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1,2,3,4,5-Pentafluoro-6-nitrobenzene
006961-5G
006961-25G
FLUO006961-5GEA
0
EUR
FLUO006961-5G
FLUO006961-25G
Pentafluornitrobenzol
Pentafluornitrobenzol
Formel:
C₆F₅NO₂ Molecular Weight: 213,06 g/mol |
MDL:
MFCD00007049 CAS-Nummer: 880-78-4 EINECS: 212-915-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...