2,3-Difluoranisol
Lieferant: FLUOROCHEM
|
Achtung
|
005734-5G
005734-1G
FLUO005734-5GEA
0
EUR
FLUO005734-5G
FLUO005734-1G
2,3-Difluoranisol
2,3-Difluoranisol
Formel:
C₇H₆F₂O Molecular Weight: 144,12 g/mol |
MDL:
MFCD01631562 CAS-Nummer: 134364-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...