2,6-Difluoranisol

Lieferant: FLUOROCHEM
Gefahr

005735-5G 005735-10G
FLUO005735-5GEA 97.2 EUR
FLUO005735-5G FLUO005735-10G
2,6-Difluoranisol
2,6-Difluoranisol
Formel: C₇H₆F₂O
Molecular Weight: 144,12 g/mol
MDL: MFCD00142846
CAS-Nummer: 437-82-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR