3,5-Difluoranisol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1,3-Difluoro-5-methoxybenzene
002350-5G
002350-25G
FLUO002350-5GEA
42.5
EUR
FLUO002350-5G
FLUO002350-25G
3,5-Difluoranisol
3,5-Difluoranisol
Formel:
C₇H₆F₂O Molecular Weight: 144,12 g/mol Siedepunkt: 145…148 °C (1013 hPa) Dichte: 1,23 g/cm³ (20 °C) |
MDL:
MFCD00042560 CAS-Nummer: 93343-10-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...