2,4,6-Trifluoranisol

Lieferant: FLUOROCHEM

Synonyme: 1-Methoxy-2,4,6-trifluorbenzol

005851-1G 005851-5G
FLUO005851-1GEA 51.8 EUR
FLUO005851-1G FLUO005851-5G
2,4,6-Trifluoranisol
2,4,6-Trifluoranisol
Formel: C₇H₅F₃O
Molecular Weight: 162.11 g/mol
MDL: MFCD01631502
CAS-Nummer: 219998-30-8
UN: 3271
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR