3,4,5-Trifluoranisol

Lieferant: FLUOROCHEM

Synonyme: 1-Methoxy-3,4,5-trifluorbenzol

005852-1G 005852-5G
FLUO005852-1GEA 48.8 EUR
FLUO005852-1G FLUO005852-5G
3,4,5-Trifluoranisol
3,4,5-Trifluoranisol
Formel: C₇H₅F₃O
Molecular Weight: 162.11 g/mol
MDL: MFCD01631451
CAS-Nummer: 203245-17-4
UN: 3271
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR