2-(Trifluormethoxy)anisol

Lieferant: CHEMPUR

Synonyme: 1-Methoxy-2-(trifluormethoxy)benzol

FL5854.1 FL5854.5 FL5854.10
CHMPFL5854.1EA 0 EUR
CHMPFL5854.1 CHMPFL5854.5 CHMPFL5854.10
2-(Trifluormethoxy)anisol
2-(Trifluormethoxy)anisol
Formel: C₈H₇F₃O₂
Molecular Weight: 192.14 g/mol
CAS-Nummer: 261952-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR