3-(Trifluormethoxy)anisol

Lieferant: FLUOROCHEM
Achtung

005855-1G 005855-5G 005855.10G
FLUO005855-1GEA 45.9 EUR
FLUO005855-1G FLUO005855-5G FLUO005855.10G
3-(Trifluormethoxy)anisol
3-(Trifluormethoxy)anisol
Formel: C₈H₇F₃O₂
MDL: MFCD01631504
CAS-Nummer: 142738-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR