2,5-Difluorphenol
Lieferant: FLUOROCHEM
001943-25G
001943-100G
FLUO001943-25GEA
0
EUR
FLUO001943-25G
FLUO001943-100G
2,5-Difluorphenol
2,5-Difluorphenol
Formel:
C₆H₄F₂O Siedepunkt: 166 °C (1013 hPa) Schmelzpunkt: 42 °C |
MDL:
MFCD00042501 CAS-Nummer: 2713-31-7 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...