2-Fluor-4-nitrophenol

Lieferant: FLUOROCHEM

002385-5G 002385-25G
FLUO002385-5GEA 0 EUR
FLUO002385-5G FLUO002385-25G
2-Fluor-4-nitrophenol
2-Fluor-4-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157.1 g/mol
Schmelzpunkt: 118…118.5 °C
MDL: MFCD00051970
CAS-Nummer: 403-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR