3-Fluor-4-nitrophenol
Lieferant: Thermo Scientific
|
Achtung
|
SB01916.1
SB01916.10
SB01916.50
MAYBSB01916.1EA
0
EUR
MAYBSB01916.1
MAYBSB01916.10
MAYBSB01916.50
3-Fluor-4-nitrophenol
3-Fluor-4-nitrophenol
Formel:
C₆H₄FNO₃ Molecular Weight: 157,1 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041251 CAS-Nummer: 394-41-2 EINECS: 206-895-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...