3-Fluor-4-nitrophenol
Lieferant: FLUOROCHEM
|
Achtung
|
001469-5G
001469-25G
FLUO001469-5GEA
48.8
EUR
FLUO001469-5G
FLUO001469-25G
3-Fluor-4-nitrophenol
3-Fluor-4-nitrophenol
Formel:
C₆H₄FNO₃ Molecular Weight: 157.1 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041251 CAS-Nummer: 394-41-2 EINECS: 206-895-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...