2,3,6-Trifluorphenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B20379.03
B20379.06
B20379.03EA
36
EUR
B20379.03
B20379.06
2,3,6-Trifluorphenol ≥98%
2,3,6-Trifluorphenol
Purity: 98%
CAS No.: 113798-74-6
Molecular Formula: C6H3F3O
Formula Weight: 148.09
Flash Point: 63°(145°F)
Melting Point: 31-36°
Beilstein Reference: 4976927
UN No.: 3261
MDL No.: MFCD00061214
Formel:
C₆H₃F₃O Molecular Weight: 148.08 g/mol Siedepunkt: 155 Schmelzpunkt: 31…36 °C Flammpunkt: 63 °C (145 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061214 CAS-Nummer: 113798-74-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...