2,4,6-Trifluorphenol

Lieferant: FLUOROCHEM

01898.5G 001898-1G 001898-5G
FLUO01898.5GEA 0 EUR
FLUO01898.5G FLUO001898-1G FLUO001898-5G
2,4,6-Trifluorphenol
2,4,6-Trifluorphenol
Formel: C₆H₃F₃O
Molecular Weight: 148,08 g/mol
Siedepunkt: 155 °C (1013 hPa)
Schmelzpunkt: 50 °C
MDL: MFCD00061216
CAS-Nummer: 2268-17-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR