3-Fluorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 3-Fluorphenylmercaptan, 3-Fluorbenzolthiol, m-Fluorthiophenol, 1-Fluor-3-mercaptobenzol, 3-Fluorbenzol-1-thiol

001299-5G 001299-25G
FLUO001299-5GEA 39.9 EUR
FLUO001299-5G FLUO001299-25G
3-Fluorthiophenol
3-Fluorthiophenol
Formel: C₆H₅FS
Molecular Weight: 128,17 g/mol
Siedepunkt: 164…166 °C (1013 hPa)
Dichte: 1,18 g/cm³ (30 °C)
MDL: MFCD00040227
CAS-Nummer: 2557-77-9
EINECS: 219-876-2
UN: 3071
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR