Iodbenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Phenyliodid

A12788.0B A12788.18 A12788.22
A12788.0BEA 472 EUR
A12788.0B A12788.18 A12788.22
Iodbenzol ≥98%
Iodbenzol

Purity: 98% CAS No.: 591-50-4 Molecular Formula: C6H5I Formula Weight: 204.01 Flash Point: 74°(165°F) Physical Form: Liquid Density: 1.826 Refractive Index: 1.6200 Sensitivity: Light Sensitive Appearance: Colourless to pale yellow Odour: Aromatic Melting Point: -31° Boiling Point: 188-189° Beilstein Reference: 1446140 Merck Reference: 14,5029 MDL No.: MFCD00001029

Formel: C₆H₅I
Molecular Weight: 204.01 g/mol
Siedepunkt: 188…189 °C
Schmelzpunkt: –31 °C
Dichte: 1.826
Flammpunkt: 74 °C (165 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001029
CAS-Nummer: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR