4-Hydroxy-3,5-diiodbenzaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Diiodo4-hydroxybenzaldehyde, 3,5-Diiodo-4-hydroxybenzaldehyde

L01060.09 L01060.18
L01060.09EA 78.8 EUR
L01060.09 L01060.18
4-Hydroxy-3,5-diiodbenzaldehyd ≥98%
4-Hydroxy-3,5-diiodbenzaldehyd

Purity: 98+% CAS No.: 1948-40-9 Molecular Formula: C7H4I2O2 Formula Weight: 373.92 Melting Point: 198-206° Beilstein Reference: 1283414

Formel: C₇H₄I₂O₂
Molecular Weight: 373,92 g/mol
Schmelzpunkt: 198…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014670
CAS-Nummer: 1948-40-9
EINECS: 217-754-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR