4-Iodbenzoesäure
Lieferant: FLUOROCHEM
Synonyme:
p-Iodobenzoic acid
005464-25G
005464-100G
FLUO005464-25GEA
48.8
EUR
FLUO005464-25G
FLUO005464-100G
4-Iodbenzoesäure
4-Iodbenzoesäure
Formel:
C₇H₅IO₂ Molecular Weight: 248,02 g/mol Siedepunkt: 319 °C (1013 hPa) Dichte: 2,184 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002533 CAS-Nummer: 619-58-9 EINECS: 210-603-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...