5-Iodsalicylsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxy-5-iodobenzoic acid

B21291.14 B21291.22 B21291.36
B21291.14EA 38.5 EUR
B21291.14 B21291.22 B21291.36
5-Iodsalicylsäure ≥97%
5-Iodsalicylsäure

Purity: 97% CAS No.: 119-30-2 Molecular Formula: C7H5IO3 Formula Weight: 264.02 Melting Point: 194-199° Beilstein Reference: 2090443

Formel: C₇H₅IO₃
Molecular Weight: 264,02 g/mol
Siedepunkt: 371 °C (1013 hPa)
Schmelzpunkt: 194…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002458
CAS-Nummer: 119-30-2
EINECS: 204-313-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR