2-Iodanisol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Iodphenolmethylether
A10150.14
A10150.22
A10150.36
A10150.14EA
40.1
EUR
A10150.14
A10150.22
A10150.36
2-Iodanisol ≥99%
2-Iodanisol
Purity: 99%
CAS No.: 529-28-2
Molecular Formula: C7H7IO
Formula Weight: 234.04
Flash Point:
Formel:
C₇H₇IO Molecular Weight: 234,04 g/mol Siedepunkt: 240…242 °C Dichte: 1,800 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001039 CAS-Nummer: 529-28-2 EINECS: 208-456-4 Merck Index: 14,5028 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...