2-Iodanisol
Lieferant: FLUOROCHEM
Synonyme:
2-Iodphenolmethylether
001302-100G
001302-25G
FLUO001302-100GEA
159
EUR
FLUO001302-100G
FLUO001302-25G
2-Iodanisol
2-Iodanisol
Formel:
C₇H₇IO Molecular Weight: 234,04 g/mol Siedepunkt: 238 °C (1013 hPa) Dichte: 1,445 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001039 CAS-Nummer: 529-28-2 EINECS: 208-456-4 Merck Index: 13,05050 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...