3-Iodanisol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-iodoanisole
A14208.06
A14208.14
A14208.06EA
29.2
EUR
A14208.06
A14208.14
3-Iodanisol ≥98%
3-Iodanisol
Purity: 98%
CAS No.: 766-85-8
Molecular Formula: C7H7IO
Formula Weight: 234.04
Flash Point:
Formel:
C₇H₇IO Molecular Weight: 234,04 g/mol Siedepunkt: 243…244 °C Dichte: 1,780 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001048 CAS-Nummer: 766-85-8 EINECS: 212-173-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...