3-Iodphenol

Lieferant: FLUOROCHEM
Achtung

001308-50G 001308-100G
FLUO001308-50GEA 0 EUR
FLUO001308-50G FLUO001308-100G
3-Iodphenol
3-Iodphenol
Formel: C₆H₅IO
Molecular Weight: 220.01 g/mol
MDL: MFCD00002261
CAS-Nummer: 626-02-8
EINECS: 210-923-2
UN: 3335
ADR: 9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR