Methylorange ACS
Lieferant: Thermo Scientific
Gefahr
Synonyme: Goldorange, Helianthin, Natrium-4-(4-dimethylaminophenylazo)benzolsulfonat, Orange III, 4-(4-(Dimethylamino)phenylazo)benzolsulfonsäure Natriumsalz, (4-(Dimethylamino)azobenzol-4'-sulfonsäure Natriumsalz
17874.06 17874.14 17874.22
17874.06EA 22.1 EUR
17874.06 17874.14 17874.22
Methylorange ACS
Methylorange
CAS No.: 547-58-0 Molecular Formula: C14H14N3NaO3S Formula Weight: 327.34 Physical Form: Crystalline Appearance: Orange Odour: Weak, characteristic pH: 6.5 Melting Point: 300° Solubility: Soluble in 500 parts water. More soluble in hot water Beilstein Reference: 4732884 Merck Reference: 14,6105 UN No.: 3143 MDL No.: MFCD00007502
Formel: C₁₄H₁₄N₃NaO₃S
Molecular Weight: 327,34 g/mol
Siedepunkt: ∼90 °C (1013 hPa)
Schmelzpunkt: >300 °C
Dichte: ∼1 g/cm³ (20 °C)
Flammpunkt: 38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007502
CAS-Nummer: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 14,6105