N,N-Dimethylanilin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Dimethylaminobenzol
A11916.0D
A11916.0F
A11916.AE
A11916.AP
A11916.0DEA
384
EUR
A11916.0D
A11916.0F
A11916.AE
A11916.AP
N,N-Dimethylanilin ≥99%
N,N-Dimethylanilin
Purity: 99%
CAS No.: 121-69-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 62°(143°F)
Physical Form: Liquid
Density: 0.956
Refractive Index: 1.5580
Odour: Characteristic
pH: 7.4
Melting Point: 2°
Boiling Point: 193°
Beilstein Reference: 507140
Merck Reference: 14,3234
UN No.: 2253
MDL No.: MFCD00008304
Formel:
C₈H₁₁N Molecular Weight: 121,18 g/mol Siedepunkt: 193 °C Schmelzpunkt: 2 °C Dichte: 0,956 Flammpunkt: 62 °C (143 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008304 CAS-Nummer: 121-69-7 EINECS: 204-493-5 UN: 2253 ADR: 6.1,II Merck Index: 14,3234 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...