N,N-Diethyl-4-nitroanilin

Lieferant: FLUOROCHEM

001750-1G 001750-5G
FLUO001750-1GEA 72.9 EUR
FLUO001750-1G FLUO001750-5G
N,N-Diethyl-4-nitroanilin
N,N-Diethyl-4-nitroanilin
Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194,23 g/mol
Schmelzpunkt: 77 °C
Dichte: 1,225 g/cm³ (20 °C)
MDL: MFCD00043606
CAS-Nummer: 2216-15-1
EINECS: 218-685-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR