N,N-Dimethyl-3-nitroanilin 98%

Lieferant: Thermo Fisher Scientific
Achtung

116470050 116470250
ACRO116470050EA 24.7 EUR
ACRO116470050 ACRO116470250
N,N-Dimethyl-3-nitroanilin 98%
N,N-Dimethyl-3-nitroanilin
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
Siedepunkt: 280…285 °C (1013 hPa)
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007236
CAS-Nummer: 619-31-8
EINECS: 210-590-3
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Orange to orange-red Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 57 to 61 °C
Titration with HClO4 ≥97.5 %
Nitrate (NO3) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR