Anilin ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Aminobenzol, Phenylamin

A14443.0B A14443.0I A14443.22 A14443.PG
A14443.0BEA 43.8 EUR
A14443.0B A14443.0I A14443.22 A14443.PG
Anilin ≥99%
Anilin

Purity: 99+% CAS No.: 62-53-3 Molecular Formula: C6H7N Formula Weight: 93.13 Flash Point: 70°(158°F) Refractive Index: 1.5860 Melting Point: -6° Boiling Point: 183-184° Solubility: Soluble in water Beilstein Reference: 605631 Merck Reference: 14,659 UN No.: 1547 MDL No.: MFCD00007629

Formel: C₆H₅NH₂
Molecular Weight: 93,13 g/mol
Siedepunkt: 183…184 °C
Schmelzpunkt: –6 °C
Dichte: 1,021
Flammpunkt: 70 °C (158 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007629
CAS-Nummer: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 14,659
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR