2-Nitroanilin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
o-Nitroanilin
128350050
128351000
128355000
128350051
ACRO128350050EA
25.4
EUR
ACRO128350050
ACRO128351000
ACRO128355000
ACRO128350051
2-Nitroanilin 98%
2-Nitroanilin
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Siedepunkt: 284 °C (760 mmHg) Schmelzpunkt: 70…74 °C Dichte: 1,446 g/cm³ (20 °C) Flammpunkt: 168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007687 CAS-Nummer: 88-74-4 EINECS: 201-855-4 UN: 1661 ADR: 6.1,II Merck Index: 12,06680 |
Spezifikation Testergebnisse
Appearance | Orange to brown Crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
GC | ≥97.5 % |
Calcium (Ca) | ≤80 ppm (Typical) |
Chromium (Cr) | ≤0.5 ppm (Typical) |
Iron (Fe) | ≤65 ppm (Typical) |
Potassium (K) | ≤50 ppm (Typical) |
Titanium (Ti) | ≤150 ppm (Typical) |
Silicon dioxide (SiO2) | ≥99.80 % (Typical) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...