2,5-Dimethoxy-4-nitroanilin
Lieferant: CHEMPUR
|
Achtung
|
BTB13704.250
BTB13704.1
CHMPBTB13704.250EA
0
EUR
CHMPBTB13704.250
MAYBBTB13704.1
2,5-Dimethoxy-4-nitroanilin
2,5-Dimethoxy-4-nitroanilin
Formel:
C₈H₁₀N₂O₄ Molecular Weight: 198,18 g/mol Schmelzpunkt: 158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007361 CAS-Nummer: 6313-37-7 EINECS: 228-640-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...