3,4,5-Trimethoxyanilin 97%
Lieferant: Thermo Scientific
|
Achtung
|
139900100
139900250
ACRO139900100EA
50.4
EUR
ACRO139900100
ACRO139900250
3,4,5-Trimethoxyanilin 97%
3,4,5-Trimethoxyanilin
Formel:
C₉H₁₃NO₃ Molecular Weight: 183,21 g/mol Siedepunkt: 293 °C (1013 hPa) Schmelzpunkt: 110…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008393 CAS-Nummer: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Off-white to beige to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 108 to 113 °C |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...