2-Nitrophenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: o-Nitrophenol

A12333.0B A12333.0I A12333.30
A12333.0BEA 82.1 EUR
A12333.0B A12333.0I A12333.30
2-Nitrophenol ≥98%
2-Nitrophenol

Purity: 98% CAS No.: 88-75-5 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Flash Point: 108°(226°F) Odour: Phenol-like Boiling Point: 214-216° Beilstein Reference: 775403 Merck Reference: 14,6620 UN No.: 1663 MDL No.: MFCD00011688

Formel: C₆H₅NO₃
Molecular Weight: 139.11 g/mol
Siedepunkt: 214…216 °C
Schmelzpunkt: 44…48 °C
Dichte: 1.26
Flammpunkt: 108 °C (226 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011688
CAS-Nummer: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1 III
Merck Index: 12,06717

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR