2-Nitrophenylphenylether

Lieferant: Thermo Scientific

Synonyme: 2-Nitrodiphenyl ether, 2-Nitrophenyl phenyl ether

CD09530.1 CD09530.50
MAYBCD09530.1EA 0 EUR
MAYBCD09530.1 MAYBCD09530.50
2-Nitrophenylphenylether
2-Nitrophenylphenylether
Formel: C₁₂H₉NO₃
Molecular Weight: 215,21 g/mol
MDL: MFCD00035744
CAS-Nummer: 2216-12-8
EINECS: 218-684-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR