Triphenylamin 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
140310250
140311000
ACRO140310250EA
66.2
EUR
ACRO140310250
ACRO140311000
Triphenylamin 99+%
Triphenylamin
Formel:
C₁₈H₁₅N Molecular Weight: 245.32 g/mol Siedepunkt: 347…348 °C (760 mmHg) Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003020 CAS-Nummer: 603-34-9 EINECS: 210-035-5 |
Spezifikation Testergebnisse
Appearance | Off-white to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 125 to 127 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...