3-Ethoxyphenol ≥98%

Lieferant: Thermo Scientific
Achtung

B24266.06 B24266.14 B24266.22
B24266.06EA 46.3 EUR
B24266.06 B24266.14 B24266.22
3-Ethoxyphenol ≥98%
3-Ethoxyphenol

Purity: 98% CAS No.: 621-34-1 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 93°(199°F) Refractive Index: 1.5410 Odour: Phenol-like Melting Point: 28-31° Boiling Point: 247° MDL No.: MFCD00016450

Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 247 °C
Schmelzpunkt: 28…31 °C
Dichte: 1,070
Flammpunkt: >93 °C (199 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016450
CAS-Nummer: 621-34-1
EINECS: 210-681-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR