Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen

Lieferant: FLUOROCHEM

Synonyme: 2-Bromotetrafluoroethyl trifluorovinyl ether

010436-25G 010436-5G
FLUO010436-25GEA 0 EUR
FLUO010436-25G FLUO010436-5G
Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen
Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen
Formel: F₂CCFOCF₂CF₂Br
Molecular Weight: 276.94 g/mol
MDL: MFCD02183517
CAS-Nummer: 85737-06-0
EINECS: 288-516-4
UN: 3295
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR