Veratrol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether
A11985.0B
A11985.0I
A11985.30
A11985.0BEA
93
EUR
A11985.0B
A11985.0I
A11985.30
Veratrol ≥99%
Veratrol
Purity: 99%
CAS No.: 91-16-7
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 87°(188°F)
Refractive Index: 1.5330
Melting Point: 21-23°
Boiling Point: 206-207°
Beilstein Reference: 1364621
Merck Reference: 14,9956
MDL No.: MFCD00008357
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 206…207 °C Schmelzpunkt: 21…23 °C Dichte: 1,084 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008357 CAS-Nummer: 91-16-7 EINECS: 202-045-3 Merck Index: 14,9956 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...