Xanthin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3,7-Dihydro-1H-purin-2,6-dion, 2,6(1H,3H)-Purindion, 1H,3H-Purin-2,6-dion, 2,6-Dihydroxypurin, 3,7-Dihydro-2H-purin-2,6-dion

X0626-10G X0626-25G X0626-5G
SIALX0626-10GEA 200 EUR
SIALX0626-10G SIALX0626-25G SIALX0626-5G
Xanthin, Sigma-Aldrich®
Xanthin
Formel: C₅H₄N₄O₂
Molecular Weight: 152.11 g/mol
MDL: MFCD00078453
CAS-Nummer: 69-89-6
EINECS: 200-718-6
Merck Index: 12,10193

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR