Ammoniumperoxodisulfat (APS), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Peroxidisulphuric acid diammonium salt, Ammonium persulphate
A3678-100G
A3678-25G
SIALA3678-100GEA
195
EUR
SIALA3678-100G
SIALA3678-25G
Ammoniumperoxodisulfat (APS), Sigma-Aldrich®
Ammoniumperoxodisulfat (APS)
Formel:
(NH₄)₂S₂O₈ Molecular Weight: 228.2 g/mol Schmelzpunkt: 120 Dichte: 1.98 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003390 CAS-Nummer: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1 III Merck Index: 13,00545 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...