2-Brom-4-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck

722413-1G 722413-5G
SIAL722413-1GEA 34.2 EUR
SIAL722413-1G SIAL722413-5G
2-Brom-4-chlorpyridin, Sigma-Aldrich®
2-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
MDL: MFCD03840757
CAS-Nummer: 22918-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR